Learn More
Selleck Chemical LLC MI-2 (Menin-MLL Inhibitor) 5mg 1271738-62-5
SDP
Supplier:  Selleck Chemical LLC S76185MG
Encompass_Preferred
MI-2 (Menin-MLL Inhibitor) is a potent menin-MLL interaction inhibitor with IC50 of 446 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8043