Learn More
Cayman Chemical MI-2hydrochlorIde 10mg
SDP
Catalog No. 502953902
Encompass_Preferred
Potently binds menin, blocks the menin-MLL fusion protein interaction (IC50 = 0.45 μM), and induces apoptosis in cells expressing MLL fusion proteins
Catalog No. 50-295-3902 Supplier Cayman Chemical Supplier No. 1162010 MG
Potently binds menin, blocks the menin-MLL fusion protein interaction (IC50 = 0.45 μM), and induces apoptosis in cells expressing MLL fusion proteins