Learn More
Selleck Chemical LLC MI-3
SDP
Catalog No. 5000004034
Encompass_Preferred
MI-3 (Menin-MLL Inhibitor) is a potent menin-MLL interaction inhibitor with IC50 of 648 nM
Catalog No. 50-000-04034 Supplier Selleck Chemical LLC Supplier No. S76195MG
MI-3 (Menin-MLL Inhibitor) is a potent menin-MLL interaction inhibitor with IC50 of 648 nM