Learn More
Medchemexpress LLC HY-15223 5mg Medchemexpress, MI-3 CAS:1271738-59-0 Purity:>98%
SDP
Catalog No. 502081694
Encompass_Preferred
Medchemexpress, HY-15223 5mg MI-3 CAS:1271738-59-0 MI-3 (Menin-MLL inhibitor 3) is a potent and high affinity menin-MLL inhibitor with an IC50 of 648 nM and a Kd of 201 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1694 Supplier Medchemexpress LLC Supplier No. HY152235MG
Medchemexpress, HY-15223 5mg MI-3 CAS:1271738-59-0 MI-3 (Menin-MLL inhibitor 3) is a potent and high affinity menin-MLL inhibitor with an IC50 of 648 nM and a Kd of 201 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.