Learn More
Medchemexpress LLC Mi-389 | 2222635-92-7 | 99.3% | 654.69 g/mol | C35H35FN6O6 | 5 MG
SDP
Catalog No. 5000222066
Encompass_Preferred
MI-389 is a proteolysis-targeting chimera (PROTAC) that degrades the translation termination factor GSPT1 through recruitment of the CRL4CRBN E3 ligase. It is provided as a research-grade small molecule with reported antiproliferative activity in leukemia cell lines and is intended for in vitro and cellular studies.
Catalog No. 50-002-22066 Supplier Medchemexpress LLC Supplier No. HY1366885MG
MI-389 is a proteolysis-targeting chimera (PROTAC) that degrades the translation termination factor GSPT1 through recruitment of the CRL4CRBN E3 ligase. It is provided as a research-grade small molecule with reported antiproliferative activity in leukemia cell lines and is intended for in vitro and cellular studies.