Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mi-389 | 2222635-92-7 | 99.3% | C35H35FN6O6 | 10MG
SDP

Supplier:  Medchemexpress LLC HY13668810MG

Encompass_Preferred

MI-389 is a PROTAC small-molecule degrader that recruits cereblon to induce proteasomal degradation of the translation termination factor GSPT1. The compound is provided for research use with defined molecular formula, characterized purity, solubility data, and storage recommendations to support reproducible in vitro and cellular studies.

  • High purity (99.3%).
  • Targets cereblon and GSPT1.
  • Soluble in DMSO; use newly opened DMSO due to hygroscopic effects.
  • Stable under recommended storage: 4°C as solid; in solution -80°C up to 6 months, -20°C up to 1 month.
  • Intended for PROTAC-focused cellular and biochemical research applications.

Catalog No. 50-002-59661


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.