Learn More
Medchemexpress LLC MI-463 | 1628317-18-9 | 99.7% | 484.54 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY19809100MG
Encompass_Preferred
MI-463 is a highly potent and orally bioavailable small molecule inhibitor of the menin-mLL interaction.
  • Highly potent and orally bioavailable.
  • Inhibits menin-mLL interaction.
  • Achieves substantial growth inhibition of mLL-AF9 transformed mouse bone marrow cells.
  • Delays progression of mLL leukemia in murine models without toxicity.
  • Provides strong inhibition of tumor growth with once daily intraperitoneal administration.
  • Significantly reduces expression of mLL fusion protein target genes, HOXA9 and MEIS1.
Catalog No. 50-004-01875