Learn More
Selleck Chemical LLC MI-463
SDP
Catalog No. 5000002879
Encompass_Preferred
MI-463 is a potent inhibitor of Menin-MLL interaction with an IC50 value of 15 3 nM
Catalog No. 50-000-02879 Supplier Selleck Chemical LLC Supplier No. S78162MG
Add to Cart
Add to Cart
MI-463 is a potent inhibitor of Menin-MLL interaction with an IC50 value of 15 3 nM