Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC MI-538 | 1857417-10-7 | 98.2% | 566.60 | 1 ML
SDP

Catalog No. 5000402009
Encompass_Preferred

MI-538 is an inhibitor of the interaction between menin and MLL fusion proteins with an IC50 of 21 nM. It is for research use only and not sold to patients.

  • Inhibits menin and MLL fusion proteins interaction
  • Potent cellular activity with improved binding affinity to menin
  • Strong down-regulation of Hoxa9 and Meis1 genes expression
  • Reduces tumor volume by approximately 80% in mice xenograft models
  • Improved exposure (AUC values), Cmax, and lowest clearance
  • Half-life: approximately 1.6 hours
  • Oral bioavailability: approximately 50%

Catalog No. 50-004-02009 Supplier Medchemexpress LLC Supplier No. HY1981010MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MI-538 is an inhibitor of the interaction between menin and MLL fusion proteins with an IC50 of 21 nM. It is for research use only and not sold to patients.

  • Inhibits menin and MLL fusion proteins interaction
  • Potent cellular activity with improved binding affinity to menin
  • Strong down-regulation of Hoxa9 and Meis1 genes expression
  • Reduces tumor volume by approximately 80% in mice xenograft models
  • Improved exposure (AUC values), Cmax, and lowest clearance
  • Half-life: approximately 1.6 hours
  • Oral bioavailability: approximately 50%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.