Learn More
Medchemexpress LLC MI-538 | 1857417-10-7 | 98.2% | 566.60 | 100 MG
SDP
Catalog No. 5000400466
Encompass_Preferred
MI-538 is an inhibitor of the interaction between menin and MLL fusion proteins with an IC50 of 21 nM. It offers selective inhibition of MLL leukemia cells and demonstrates efficacy in vivo.
  • Inhibits proliferation of MLL leukemia cells
  • Binds to menin with low nanomolar affinity
  • Downregulates Hoxa9 and Meis1 gene expression
  • Reduces MV4;11 tumor volume in mice xenografts
  • Exhibits high oral bioavailability
Catalog No. 50-004-00466 Supplier Medchemexpress LLC Supplier No. HY19810100MG
MI-538 is an inhibitor of the interaction between menin and MLL fusion proteins with an IC50 of 21 nM. It offers selective inhibition of MLL leukemia cells and demonstrates efficacy in vivo.
  • Inhibits proliferation of MLL leukemia cells
  • Binds to menin with low nanomolar affinity
  • Downregulates Hoxa9 and Meis1 gene expression
  • Reduces MV4;11 tumor volume in mice xenografts
  • Exhibits high oral bioavailability