Learn More
Cayman Chemical MI-nchydrochlorIde 10mg
SDP
Catalog No. 502952525
Encompass_Preferred
A weak inhibitor of the menin-MLL fusion protein interaction (IC50 = 193 μM), intended as a negative control compound for tests involving MI-2
Catalog No. 50-295-2525 Supplier Cayman Chemical Supplier No. 1162110 MG
A weak inhibitor of the menin-MLL fusion protein interaction (IC50 = 193 μM), intended as a negative control compound for tests involving MI-2