Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC MIA Human N-His 10ug
SDP

Catalog No. 5000197009
Encompass_Preferred

MIA Protein, Human (His) is a recombinant form of human Melanoma Inhibitory Activity (MIA) expressed in Escherichia coli and supplied with a His tag. It is intended for research applications to investigate MIA's effects on melanoma cell growth and its protein-protein interactions in biochemical and cellular assays. The product corresponds to UniProt Q16674 (residues G25-Q131) and is provided at research-scale quantities suitable for assay development.

  • Recombinant human MIA expressed in Escherichia coli.
  • C-6*His tag for purification and detection.
  • Corresponds to residues G25-Q131 of UniProt Q16674.
  • Predicted molecular mass ~13.3 kDa (≈14 kDa by SDS-PAGE).
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Low endotoxin level (<1 EU/μg) suitable for cell-based assays.
  • Supplied in research-scale quantities for biochemical assays.

Catalog No. 50-001-97009 Supplier Medchemexpress LLC Supplier No. HYP70846A10UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MIA Protein, Human (His) is a recombinant form of human Melanoma Inhibitory Activity (MIA) expressed in Escherichia coli and supplied with a His tag. It is intended for research applications to investigate MIA's effects on melanoma cell growth and its protein-protein interactions in biochemical and cellular assays. The product corresponds to UniProt Q16674 (residues G25-Q131) and is provided at research-scale quantities suitable for assay development.

  • Recombinant human MIA expressed in Escherichia coli.
  • C-6*His tag for purification and detection.
  • Corresponds to residues G25-Q131 of UniProt Q16674.
  • Predicted molecular mass ~13.3 kDa (≈14 kDa by SDS-PAGE).
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Low endotoxin level (<1 EU/μg) suitable for cell-based assays.
  • Supplied in research-scale quantities for biochemical assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.