Learn More
ATCC MIA PaCa-2; Pancreatic Carcinoma; Human (Homo sapiens)
Supplier:  ATCC CRL1420
Encompass
MIA PaCa-2 is an epithelial cell line that was derived from tumor tissue of the pancreas obtained from a 65-year-old, White male. This cell line can be used in cancer research.
Catalog No. 50-238-5009