Learn More
ATCC MIA PaCa-2; Pancreatic Carcinoma; Human (Homo sapiens)
Catalog No. 502384590
Encompass
MIA PaCa-2 is an epithelial cell that was isolated from the pancreas of a 65-year-old, White male with carcinoma. The cell line can be used in testing and calibration in ISO 17025-accredited laboratories, to challenge assay performance, validate or compare test methods, and establish sensitivity, linearity, and specificity during assay validation or implementation.
Catalog No. 50-238-4590 Supplier ATCC Supplier No. CRMCRL1420
MIA PaCa-2 is an epithelial cell that was isolated from the pancreas of a 65-year-old, White male with carcinoma. The cell line can be used in testing and calibration in ISO 17025-accredited laboratories, to challenge assay performance, validate or compare test methods, and establish sensitivity, linearity, and specificity during assay validation or implementation.