Learn More
Aviva Systems Biology Corp MIA2 Antibody - C-terminal region (ARP60753_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60753_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MIA2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human MIA2. Alternate names: FLJ22404ApplicationWB. Species reactivity: Human
Catalog No. 50-287-147