Learn More
Medchemexpress LLC Mianserin | 24219-97-4 | 99.42% | 25 MG
SDP
Catalog No. 5000450660
Encompass_Preferred
Mianserin is an orally active H1 receptor antagonist that can also activate κ-opioid and octopamine receptors. It increases ERK1/2 and CREB phosphorylation and antagonizes full κ-opioid agonist and Dynorphin A-induced MAPK phosphorylation. Mianserin modulates social and exploratory behavior and raises electroconvulsive thresholds. It is used in the research of neurological diseases such as depression and epilepsy.
  • Orally active H1 receptor antagonist
  • Activates κ-opioid receptor and octopamine receptor
  • Increases ERK1/2 and CREB phosphorylation
  • Antagonizes full κ-opioid agonist and Dynorphin A-induced MAPK phosphorylation
  • Modulates social and exploratory behavior
  • Raises electroconvulsive thresholds
  • Useful for research into neurological diseases like depression and epilepsy
Catalog No. 50-004-50660 Supplier Medchemexpress LLC Supplier No. 25MGHYB0188
Mianserin is an orally active H1 receptor antagonist that can also activate κ-opioid and octopamine receptors. It increases ERK1/2 and CREB phosphorylation and antagonizes full κ-opioid agonist and Dynorphin A-induced MAPK phosphorylation. Mianserin modulates social and exploratory behavior and raises electroconvulsive thresholds. It is used in the research of neurological diseases such as depression and epilepsy.
  • Orally active H1 receptor antagonist
  • Activates κ-opioid receptor and octopamine receptor
  • Increases ERK1/2 and CREB phosphorylation
  • Antagonizes full κ-opioid agonist and Dynorphin A-induced MAPK phosphorylation
  • Modulates social and exploratory behavior
  • Raises electroconvulsive thresholds
  • Useful for research into neurological diseases like depression and epilepsy