Learn More
Medchemexpress LLC Mianserin-d3 hydrochloride | 1219804-97-3 | 97.51% | 1 MG
SDP
Catalog No. 5000405954
Encompass_Preferred
Mianserin-d3 hydrochloride is a deuterium-labeled form of Mianserin, an orally active H1 receptor antagonist that can also activate κ-opioid and octopamine receptors. It increases ERK1/2 and CREB phosphorylation and antagonizes full κ-opioid agonist and Dynorphin A-induced MAPK phosphorylation. This compound has been observed to modulate social and exploratory behavior and raise electroconvulsive thresholds. It is investigated for research into neurological diseases like depression and epilepsy. Stable heavy isotopes, like deuterium, are incorporated into drug molecules for quantitation during drug development and can influence pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-05954 Supplier Medchemexpress LLC Supplier No. HYB0188AS1MG
Mianserin-d3 hydrochloride is a deuterium-labeled form of Mianserin, an orally active H1 receptor antagonist that can also activate κ-opioid and octopamine receptors. It increases ERK1/2 and CREB phosphorylation and antagonizes full κ-opioid agonist and Dynorphin A-induced MAPK phosphorylation. This compound has been observed to modulate social and exploratory behavior and raise electroconvulsive thresholds. It is investigated for research into neurological diseases like depression and epilepsy. Stable heavy isotopes, like deuterium, are incorporated into drug molecules for quantitation during drug development and can influence pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS