Learn More
TARGETMOL CHEMICALS INC MIBAMPATOR 5MG
SDP
Catalog No. 502581199
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Mibampator (LY451395) is an effective and selective AMPA receptor potentiator. purity: 97%
Catalog No. 50-258-1199 Supplier TARGETMOL CHEMICALS INC Supplier No. T158205MG
Also available in 1 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Mibampator (LY451395) is an effective and selective AMPA receptor potentiator. purity: 97%