Learn More
Medchemexpress LLC Mibampator (LY451395) | 375345-95-2 | 99.5% | 438.60 | 100 MG
SDP
Catalog No. 5000345198
Encompass_Preferred
Mibampator (LY451395) is a potent and highly selective potentiator of the AMPA receptors, intended for research use only. It has shown various cellular effects, including toxicity against Caco-2 cells and inhibition of SARS-CoV-2 induced cytotoxicity. In vivo studies indicated its ability to reverse ethanol-induced deficits in rats. It has also undergone Phase 2 clinical trials for Alzheimer's Disease.
  • Potent and highly selective potentiator of AMPA receptors
  • Caco-2 CC50 of 8.66 μM and IC50 of 3.28 μM
  • HEK293 EC50 of 0.15 μM
  • Reverses central effects of acutely intoxicating dose of ethanol in rats
  • Clinical trials for Alzheimer's Disease
Catalog No. 50-003-45198 Supplier Medchemexpress LLC Supplier No. HY1093410MG
Mibampator (LY451395) is a potent and highly selective potentiator of the AMPA receptors, intended for research use only. It has shown various cellular effects, including toxicity against Caco-2 cells and inhibition of SARS-CoV-2 induced cytotoxicity. In vivo studies indicated its ability to reverse ethanol-induced deficits in rats. It has also undergone Phase 2 clinical trials for Alzheimer's Disease.
  • Potent and highly selective potentiator of AMPA receptors
  • Caco-2 CC50 of 8.66 μM and IC50 of 3.28 μM
  • HEK293 EC50 of 0.15 μM
  • Reverses central effects of acutely intoxicating dose of ethanol in rats
  • Clinical trials for Alzheimer's Disease