Learn More
Medchemexpress LLC HY-15553A 5mg , Mibefradil (dihydrochloride) CAS:116666-63-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY15553A5MG
Encompass_Preferred
HY-15553A 5mg Mibefradil (dihydrochloride) CAS: 116666-63-8 Mibefradil dihydrochloride (Ro 40-5967 dihydrochloride) is a calcium channel blocker with moderate selectivity for T-type Ca2+ channels (IC50s of 2.7 μM and 18.6 μM for T-type and L-type currents, respectively). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0921