Learn More
Medchemexpress LLC HY-15553A 10mg Medchemexpress, Mibefradil (dihydrochloride) CAS:116666-63-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY15553A10MG
Encompass_Preferred
Medchemexpress, HY-15553A 10mg Mibefradil (dihydrochloride) CAS:116666-63-8 Mibefradil dihydrochloride (Ro 40-5967 dihydrochloride) is a calcium channel blocker with moderate selectivity for T-type Ca2+ channels displaying IC50s of 2.7 μM and 18.6 μM for T-type and L-type currents, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9138