Learn More
Selleck Chemical LLC Mibefradil dihydrochloride-1MG
SDP
Catalog No. 503137184
Encompass_Preferred
Mibefradil dihydrochloride (Ro 40-5967 dihydrochloride) is a calcium channel blocker with moderate selectivity for T-type Ca2 channels (IC50s of 2 7 M and 18 6 M for T-type and L-type currents respectively)
Catalog No. 50-313-7184 Supplier Selleck Chemical LLC Supplier No. E17051MG
Mibefradil dihydrochloride (Ro 40-5967 dihydrochloride) is a calcium channel blocker with moderate selectivity for T-type Ca2 channels (IC50s of 2 7 M and 18 6 M for T-type and L-type currents respectively)