Learn More
Cayman Chemical MIbefradIlhydrochlorIde 10mg
SDP
Catalog No. 502972574
Encompass_Preferred
A general calcium channel blocker that shows modest selectivity for T-type channels over L-type channels (IC50 = 2.7 and 18.6 μM, respectively); a potent vasodilator, increasing coronary artery flow with an EC50 value of 54 nM
Catalog No. 50-297-2574 Supplier Cayman Chemical Supplier No. 1503710 MG
A general calcium channel blocker that shows modest selectivity for T-type channels over L-type channels (IC50 = 2.7 and 18.6 μM, respectively); a potent vasodilator, increasing coronary artery flow with an EC50 value of 54 nM