Learn More
Roboz Surgical MIC DISSECT.SCISSOR 3.5 STRAIG
SDP
Catalog No. NC0143758
Encompass
These micro dissecting scissors are precision-engineered for delicate tissue work, featuring a straight design with sharp, pointed tips and a 3 1/2-inch overall length. The instrument utilizes specialized blades, where one blade is micro-serrated to grip tissue and prevent slippage, while the other is honed to a razor-sharp bevel to ensure clean, smooth cuts without crushing. Designed specifically for soft tissue applications, these scissors provide superior cutting performance with reduced force requirements.
  • Features one micro-serrated blade for secure tissue gripping
  • Utilizes a bevel-honed edge for sharp, non-crushing cuts
  • Constructed with a 23mm blade length and sharp, pointed tips
  • Designed exclusively for tissue dissection; not for use on bone, wire, or other hard materials
  • Straight configuration with matte grey finger rings and a black screw
Catalog No. NC0143758 Supplier Roboz Surgical Supplier No. RS5910SC
These micro dissecting scissors are precision-engineered for delicate tissue work, featuring a straight design with sharp, pointed tips and a 3 1/2-inch overall length. The instrument utilizes specialized blades, where one blade is micro-serrated to grip tissue and prevent slippage, while the other is honed to a razor-sharp bevel to ensure clean, smooth cuts without crushing. Designed specifically for soft tissue applications, these scissors provide superior cutting performance with reduced force requirements.
  • Features one micro-serrated blade for secure tissue gripping
  • Utilizes a bevel-honed edge for sharp, non-crushing cuts
  • Constructed with a 23mm blade length and sharp, pointed tips
  • Designed exclusively for tissue dissection; not for use on bone, wire, or other hard materials
  • Straight configuration with matte grey finger rings and a black screw