Learn More
Santa Cruz Biotechnology MICA Antibody (2C10), 200 ug/mL
Supplier:  Santa Cruz Biotechnology SC23870
Encompass
MICA Antibody (2C10) is a high quality monoclonal MICA antibody (also designated MICA antibody) suitable for the detection of the MICA protein of human origin. MICA Antibody (2C10) is available as both the non-conjugated anti-MICA antibody form, as well as multiple conjugated forms of anti-MICA antibody, including agarose, HRP, PE, FITC and multiple Alexa Fluor conjugates.
Catalog No. NC1764562