Learn More
Sino Biological MICA Antibody, Mouse MAb 100uL
Supplier:  Sino Biological 12302MM04100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human MICA (rh MICA; Catalog#12302-H08H; AAH16929.1; Met1-Gln308). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-224-7826