Learn More
Sino Biological MICA Antibody (PE), Mouse MAb 100Tests
Supplier:  Sino Biological 12302MM04P100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human MICA (rh MICA; Catalog#12302-H08H; AAH16929.1; Met 1-Gln 308) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-224-8228