Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human MICA ELISA Kit

Catalog No. BMS2302
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BMS2302 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BMS2302 Supplier Invitrogen™ Supplier No. BMS2302
Add to Cart
Add to Cart

ELISA

The Human MICA ELISA quantitates Hu MICA in human serum, plasma, cell lysates, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu MICA.

Principle of the method

The Human MICA solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, binding to the target on a different epitope from the capture antibody. A conjugated enzyme has been incorporated into the assay. After incubation and washing steps to rid the microplate of unbound substances, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.

Rigorous validation

Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Major histocompatibility complex (MHC) class I proteins are ubiquitously expressed and mediate the recognition of intracellular antigens by cytotoxic T cells. A related family, termed the MHC class I chain-related (MIC) proteins are recognized by NKG2D, a receptor on NK and T cells, and promote anti-tumor activity. MICA, a member of the MIC family, is widely expressed on many tumors, and it is the MICA/NKG2D interaction that is thought to stimulate the anti-tumor reactivity by T lymphocytes. Both MICA and MICB mRNA are widely expressed in normal tissues, with MICA being present in virtually every tissue except the nervous system, suggesting that MIC protein expression may only be one component of the anti-tumor activity of the immune system.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q29983
Assay Range 1.6-100 pg/mL
Assay Sensitivity 1.09 pg/mL
Content And Storage Store in refrigerator (2–8°C).
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Growth Factors & Receptors
Sample Type Cell Lysates, Plasma, Serum, Supernatant
Substrate Type Adaptor Proteins
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias DAMA-345G11.2; HLA class I antigen; MHC class I chain-related protein A; MHC class I polypeptide-related sequence A; MHC class I related sequence A; MICA; MIC-A; PERB11.1; stress inducible class I homolog; truncated MHC class I polypeptide-related sequence ADAMA-345G11.2; HLA class I antigen; MHC class I chain-related protein A; MHC class I polypeptide-related sequence A; MHC class I related sequence A; MICA; MIC-A; PERB11.1; stress inducible class I homolog; truncated MHC class I polypeptide-related sequence A;
Gene ID (Entrez) 100507436
Gene Symbol MICA
Interassay CV 6.6%
Intraassay CV 5.1%
Kit Contents Pre-coated 96 well plate, Standard (2 vials), Standard Dilution Buffer, Biotinylated Detection Antibody, Streptavidin-HRP, HRP Diluent, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard (2 vials), Standard Dilution Buffer, Biotinylated Detection Antibody, Streptavidin-HRP, HRP Diluent, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Protein Aliases MHC class I polypeptide-related sequence A, MIC-A
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Cell Lysate 50 μL, Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet Ice
Storage Requirements 2-8°C
Target MICA
Test Time 1 hr 20 min
Total Assay Time 4 hr
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.