Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mica, Water Ground, 325 Mesh, Spectrum™ Chemical
SDP

Catalog No. 18604578 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
6 x 500 g
4 x 2.5 kg
45 kg
Packaging:
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-578 45 kg Poly Pail
18-604-574 500 g Poly Pail
18-604-575 6 x 500 g Poly Pail
18-604-577 4 x 2.5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-604-578 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M128945KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

M1289, 12001-26-2

Spectrum™ Chemical Mica, Water Ground, 325 Mesh is a wet-ground mica, a type of highly brittle silicate minerals with diverse chemical composition, that retains the brilliancy of its cleavage faces. It is used in cosmetic and personal care products as a colorant with opacifying properties. Its reflective and refractive properties make mica an important ingredient in makeup, moisturizing lotions, toothpastes, and nail polish. It appears as a clear, lustrous, crystalline solid plates and is insoluble in cold water.

Chemical Identifiers

CAS 12001-26-2
Molecular Weight (g/mol) 797 g/mol

Specifications

Packaging Poly Pail
CAS Max % 100%
Quantity 45 kg
Grade Reagent
Chemical Name or Material Mica
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.