Learn More
Apexbio Technology LLC Micafungin sodium 208538-73-2 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062878
Encompass_Preferred
Micafungin sodium is an antifungal agent that acts by inhibiting the synthesis of 1 3- -D-glucan an essential polymer for fungal cell wall integrity particularly in Candida albicans In addition to antifungal activities micafungin has been studied for its capacity to suppress biofilm formation by Pseudomonas aeruginosa via downregulation of genes encoding structural biofilm elements such as algC and pelC and the 1 3- -D-glucan-encoding gene ndvB Micafungin has demonstrated synergy with other antifungal compounds in animal models in murine models of systemic Aspergillus fumigatus infection micafungin administration resulted in increased survival rates Micafungin is utilized in biomedical research to investigate antifungal mechanisms combination therapies and inhibition of pathogen biofilms Reported inhibitory activity (IC50) for fungal glucan synthase typically falls within the nanomolar range
Catalog No. 50-000-62878 Supplier Apexbio Technology LLC Supplier No. A36065.1
Micafungin sodium is an antifungal agent that acts by inhibiting the synthesis of 1 3- -D-glucan an essential polymer for fungal cell wall integrity particularly in Candida albicans In addition to antifungal activities micafungin has been studied for its capacity to suppress biofilm formation by Pseudomonas aeruginosa via downregulation of genes encoding structural biofilm elements such as algC and pelC and the 1 3- -D-glucan-encoding gene ndvB Micafungin has demonstrated synergy with other antifungal compounds in animal models in murine models of systemic Aspergillus fumigatus infection micafungin administration resulted in increased survival rates Micafungin is utilized in biomedical research to investigate antifungal mechanisms combination therapies and inhibition of pathogen biofilms Reported inhibitory activity (IC50) for fungal glucan synthase typically falls within the nanomolar range