Learn More
ACROBiosystems Recombinant Protein;Human MICA Protein;HEK293;100UG;MIA,MICA,MIC-A
Supplier:  ACROBiosystems MIAH5221100UG
Encompass_Preferred
Recombinant Protein;Human MICA Protein;100UG;Human MICA, His Tag (MIA-H5221) is expressed from human 293 cells (HEK293). It contains AA Glu 24 - Gln 308 (Accession # NP_000238).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 33.6 kDa. The protein migrates as 45-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8400