Learn More
Aviva Systems Biology Corp MICAL1 Antibody - C-terminal region (ARP61957_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61957_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MICAL1. It was validated on Western Blot. . Alternate names: DKFZp434B1517, FLJ11937, FLJ21739, MICAL, MICAL-1, NICALApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Pig, Rabbit, Rat
Catalog No. 50-287-519