Learn More
ABclonal Technology MICAL1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A15505500UL
Encompass_Preferred
This gene encodes an enzyme that oxidizes methionine residues on actin, thereby promoting depolymerization of actin filaments. This protein interacts with and regulates signalling by NEDD9/CAS-L (neural precursor cell expressed, developmentally down-regulated 9). Alternative splicing results in multiple transcript variants.
Catalog No. 50-001-05939