Learn More
Aviva Systems Biology Corp MICALL1 Antibody - N-terminal region (ARP51692_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51692_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MICALL1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MICALL1. Alternate names: DKFZp686M2226, FLJ45921, KIAA1668, MICAL-L1, MIRAB13ApplicationIHC, WB. Species reactivity: Goat, Human
Catalog No. 50-014-60587