Learn More
Medchemexpress LLC MICB Human HEK293 100ug
SDP
Catalog No. 5000180428
Encompass_Preferred
MICB Protein a heavily glycosylated ligand for NKG2D receptor triggers cytolytic response in immune cells It resembles MHC class I molecules but lacks association with beta-2-microglobulin or peptide binding Alternative splicing leads to multiple transcript variants MICB is widely expressed especially in lymph nodes and bone marrow highlighting its immune response role in diverse physiological contexts MICB Protein Human (HEK293 His) is the recombinant human-derived MICB protein expressed by HEK293 with C-His C-6 His labeled tag
Catalog No. 50-001-80428 Supplier Medchemexpress LLC Supplier No. HYP73294100UG
MICB Protein a heavily glycosylated ligand for NKG2D receptor triggers cytolytic response in immune cells It resembles MHC class I molecules but lacks association with beta-2-microglobulin or peptide binding Alternative splicing leads to multiple transcript variants MICB is widely expressed especially in lymph nodes and bone marrow highlighting its immune response role in diverse physiological contexts MICB Protein Human (HEK293 His) is the recombinant human-derived MICB protein expressed by HEK293 with C-His C-6 His labeled tag