Learn More
Braintree Scientific Mice Lab Std Gas Anesthesia Mask for 51500 series only
SDP
Catalog No. 501954867
Encompass
The Gas Anesthesia mask is installed over the incisor bar, with the cone opening toward the open U. The animal (rat or mouse) is installed by placing its teeth over the incisor bar, and sliding the adaptor forward until the cone is snug about the animals nose, thereby substituting for the nose clamp. The teeth and the cone provide a snug, stable hold. Anesthesia Masks enable gas anesthesia of rats or mice in a Lab Standard™ or other stereotaxic instruments, with minimal researcher exposure to anesthetic gases. Anesthetic gas flows in the tubing connector, past the animals nose, and out the other side. Spent gas may be routed to recycling, exhaust or gas analysis instruments. The Rat Gas Anesthesia Mask (51610) fits easily onto the 51621 Rat Adaptor (sold separately) **Sold for research purposes only and not for human use**
Catalog No. 50-195-4867 Supplier Braintree Scientific Supplier No. GM51609
The Gas Anesthesia mask is installed over the incisor bar, with the cone opening toward the open U. The animal (rat or mouse) is installed by placing its teeth over the incisor bar, and sliding the adaptor forward until the cone is snug about the animals nose, thereby substituting for the nose clamp. The teeth and the cone provide a snug, stable hold. Anesthesia Masks enable gas anesthesia of rats or mice in a Lab Standard™ or other stereotaxic instruments, with minimal researcher exposure to anesthetic gases. Anesthetic gas flows in the tubing connector, past the animals nose, and out the other side. Spent gas may be routed to recycling, exhaust or gas analysis instruments. The Rat Gas Anesthesia Mask (51610) fits easily onto the 51621 Rat Adaptor (sold separately) **Sold for research purposes only and not for human use**