Learn More
MSC Rubbermaid Dust Pad, Microfiber, 18 in, Fringed, Green
Catalog No. 502835711
Encompass
This Light Commercial Microfiber Dust Pad features fringe, providing a greater surface area for increased dust, dirt, and debris removal, leading to cleaner and healthier facilities. Its high-quality split microfiber penetrates deeper for a superior clean, and the fringe enhances debris pick-up.
  • High-quality split microfiber penetrates deeper for a superior clean.
  • Fringe increases surface area for improved debris pick-up.
  • Durable hook and loop backing securely attaches to the frame and will not degrade during commercial laundering.
  • Tested to withstand up to 200 commercial launderings, and 100 launderings with bleach.
  • Designed for use with the Light Commercial Quick-Connect Frame and Handle.
Catalog No. 50-283-5711 Supplier MSC Supplier No. 57804387
This Light Commercial Microfiber Dust Pad features fringe, providing a greater surface area for increased dust, dirt, and debris removal, leading to cleaner and healthier facilities. Its high-quality split microfiber penetrates deeper for a superior clean, and the fringe enhances debris pick-up.
  • High-quality split microfiber penetrates deeper for a superior clean.
  • Fringe increases surface area for improved debris pick-up.
  • Durable hook and loop backing securely attaches to the frame and will not degrade during commercial laundering.
  • Tested to withstand up to 200 commercial launderings, and 100 launderings with bleach.
  • Designed for use with the Light Commercial Quick-Connect Frame and Handle.