Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MichClone 10% Neutral Buffered Formalin

Catalog No. MHK425
Change view
Click to view available options
Quantity:
60 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MHK425 60 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MHK425 Supplier MichClone Associates, Inc. Supplier No. K425
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Formalin, MichClone 10% Neutral Buffered; Contains 3.7% formaldehyde with methanol as stabilizer, .4% sodium phosphate monobasic, and .65% sodium phosphate dibasic in water; Store at 20-30°C; 144/case

Specifications

Quantity 60 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.