Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Great Lakes Diagnostics MichClone Associates B-5 Fixative

Catalog No. p-2727203
Change view
Click to view available options
Quantity:
30 mL, 72/Case
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MH-K430A 30 mL, 72/Case
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MH-K430A Supplier Great Lakes Diagnostics Supplier No. K430A
Only null left
Add to Cart
Add to Cart

Includes

Prefilled vials of B-5 and 37% formaldehyde in a plastic bag; case contains 72 kits.

For use in immunohistology particularly for bone marrow, lymph nodes and spleen preparations

  • The traditional method for the fixation of leukemia and lymphoma tissues
  • Contains the pre-measured B-5 Fixative with a vial of 37% Formaldehyde
  • Prior to collection of specimen, the B-5 Fixative Kit can be mixed with minimal exposure to the histotechnologist

Specifications

Type B-5 Fixative Kit
Formulation Mercuric Chloride 12g, Sodium Acetate 2.5g, Distilled Water 200mL
Includes Prefilled vials of B-5 and 37% Formaldehyde in a Plastic bag
Quantity 30 mL, 72/Case
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.