Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Great Lakes Diagnostics MichClone Modified PVA Fixative

Catalog No. mhk504a
Change view
Click to view available options
Quantity:
100/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MH-K504A 100/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MH-K504A Supplier Great Lakes Diagnostics Supplier No. K504A
Only null left
Add to Cart
Add to Cart

Includes

30mL jars with 15mL fill, 100 sets of two test tubes

Cupric sulfate-modified PVA and neutral buffered formalin

  • Can be used to preserve fecal specimens for all intestinal parasites, exam by wet mount, concentration and permanent stain procedures
  • 10mL liquid: 31% alcohol, 5% glacial acetic acid, 5% PVA; 2% cupric sulfate, 2% glycerol, and water
  • Fecal tubes used for detecting ova and parasites when present in very low numbers

Specifications

Product Type Modified PVA Fixatve
Format Liquid
Media Type 31% Alcohol, 5% Glacial Acetic Acid, 5% PVA, 2% Cupric Sulfate, 2% Glycerol and Water
Quantity 100/Cs.
Storage Requirements 20°C to 25°C
Volume (English) 0.33 oz.
Volume (Metric) 10mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.