Learn More
Great Lakes Diagnostics MichClone Sickle Cell Solubility Buffer
Catalog No. MHK111A
Change view
Click to view available options
Quantity:
Each
Catalog No. Quantity
MH-K111A Each
1 options
Catalog No. MH-K111A Supplier Great Lakes Diagnostics Supplier No. GLD100AF
Can be used to detect the sickle cell trait (AS) of sickle cell disease (SS) hemoglobin as well as other sickling hemoglobins
  • Must perform electrophoresis and confirm positive blood to distinguish betwen SS and AS
  • For in vitro diagnostic use
  • Contains .97M potassium phospate monobasic and 1.33M potassium phosphate dibasic with thimerosal as perservative
  • Refrigerate between 2 to 8°C after reconstitution
Specifications
Type Sickle Cell Solubility Kit
Includes 0.97M Potassium Phospate Monobasic and 1.33M Potassium Phosphate Dibasic with Thimerosal as preservative
Quantity Each