Learn More
Electron Microscopy Sciences Michel's Buffer 1 LT
SDP
Catalog No. 5032031
Encompass_Preferred
Our buffers are ideal for maintaining a constant pH during fixation and have an ionic constitution to prevent the extraction of cellular components or the deposition of precipitates of fixative chemicals. The buffers are prepared fresh for each order and conveniently packaged.
Catalog No. 50-320-31 Supplier Electron Microscopy Sciences Supplier No. 1804110
Our buffers are ideal for maintaining a constant pH during fixation and have an ionic constitution to prevent the extraction of cellular components or the deposition of precipitates of fixative chemicals. The buffers are prepared fresh for each order and conveniently packaged.