Learn More
Electron Microscopy Sciences Michel's Buffer 500 ML
SDP
Catalog No. 5032030
Encompass_Preferred
Our buffers are ideal for maintaining a constant pH during fixation and have an ionic constitution to prevent the extraction of cellular components or the deposition of precipitates of fixative chemicals. The buffers are prepared fresh for each order and conveniently packaged.
Catalog No. 50-320-30 Supplier Electron Microscopy Sciences Supplier No. 1804105
Our buffers are ideal for maintaining a constant pH during fixation and have an ionic constitution to prevent the extraction of cellular components or the deposition of precipitates of fixative chemicals. The buffers are prepared fresh for each order and conveniently packaged.