Learn More
Electron Microscopy Sciences Michel's Fixative in Pre-filled Vials, pH 7.0 – 7.4 10 ML
SDP
Supplier:  Electron Microscopy Sciences 1804020
Encompass_Preferred
Michel’s Fixative is prepared fresh upon receipt of order to ensure maximum shelf life. Filled in 20 ml glass vials (10 ml/vial) allowing for fixation of your specimens without transfer.
Catalog No. 50-320-33