Learn More
Michel's Fixative 5.2 mL with Green Cap, Cenmed™

Description
- Precisely measured 5.2 mL volume and distinct green cap signify a commitment to accuracy in preserving tissue integrity
- Stand out in your histological procedures with Michel's Fixative, a trusted choice for researchers and professionals seeking unparalleled precision in tissue preservation
- Elevate your histological work with confidence, knowing your dedication to achieving excellence in every preservation step
Specifications
Specifications
| Physical Form | Liquid |
| CAS | 7783-20-2 |
| Color | Clear |
| Quantity | 500 x 5.2 mL |
| Grade | Reagent |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.