Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Michel's Fixative 5.2 mL with Red Cap, Cenmed™
SDP

Catalog No. 21520243
Change view
Click to view available options
Quantity:
500 x 5.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-520-243 500 x 5.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-520-243 Supplier Cen-Med Enterprises Supplier No. CENMED 3537855RE
Add to Cart
Add to Cart

Thoughtfully crafted with a red cap for specialized tissue preservation, Michel's Fixative is meticulously designed and guarantees precise fixation of tissue samples, setting a new benchmark for histological applications.

  • 5.2 mL volume, precisely measured, combined with the distinctive red cap, underscores a commitment to accuracy in tissue preservation
  • Stand out in histological procedures with Michel's Fixative, a trusted choice for researchers and professionals dedicated to achieving ideal tissue preservation
  • Elevate your histological work confidently, knowing that Michel's Fixative with the red cap is tailored for specialized applications, ensuring unmatched accuracy in every preservation process

Specifications

Physical Form Liquid
CAS 7783-20-2
Color Clear
Quantity 500 x 5.2 mL
Grade Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.