Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Michel's Fixative with Red Cap Glutaraldehyde 3% in 0.1 Phosphate Buffer, Cenmed™
SDP

Catalog No. 21520238
Change view
Click to view available options
Sterility:
Sterile
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility
21-520-238 Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-520-238 Supplier Cen-Med Enterprises Supplier No. REN002
Add to Cart
Add to Cart

Michel's Fixative with distinctive red cap ensures optimal tissue fixation, while Glutaraldehyde 3% in 0.1 Phosphate Buffer provides a reliable option for advanced preservation needs.

  • Enhance your laboratory procedures with specialized solutions tailored for precise specimen preservation
  • Both solutions stored in 5.2 ml vial
  • Safely store and transport your specimens using our clear-orange biohazard specimen bags, measuring 6 x 9 inches and featuring indicators for added safety assurance
  • Trust in our comprehensive solutions to maintain the integrity of your specimens, ensuring accurate results and advancing your research endeavors
  • Adding 10% Formalin offers buffered preservation, ensuring the stability of specimens for accurate analysis and research purposes and completing this set

Specifications

Product Type Specimen Collection Kit
Format Tube
Media Type Molecular Transport Media
Sterility Sterile
For Use With (Application) Collection, Transport, and Preservation of Biological Specimens
Material Polypropylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.