Learn More
StatLab Michel's Wash Solution

Description
- Used to rinse and remove Michel's Transport Medium that contains ammonium sulfate from fresh tissue specimens prior to frozen sectioning and immunofluorescence microscopy
- Clears residual transport medium components from unfixed tissue samples (such as skin or renal biopsies) so they do not interfere with subsequent processing and staining
Specifications
Specifications
| Physical Form | Solution |
| Color | Clear Colorless |
| Quantity | 1 gal. |
| Grade | Laboratory |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.