Learn More
Newcomer Supply Michel Wash Solution, 1 Liter
SDP
Catalog No. NC0229814
Encompass
Newcomer Supply Michel Wash Solution is used to rinse Michel Transport Medium from tissue after transport or storage and prior to freezing. Shelf Life is 1 year from date of manufacture. Store at 15-30°C.
Catalog No. NC0229814 Supplier Newcomer Supply Supplier No. 1243B
Newcomer Supply Michel Wash Solution is used to rinse Michel Transport Medium from tissue after transport or storage and prior to freezing. Shelf Life is 1 year from date of manufacture. Store at 15-30°C.