Learn More
Newcomer Supply Michel Transport Medium, 500 ml. Preserves morphology & antigenicity during transport of tissues
SDP
Catalog No. NC0777939
Encompass
Newcomer Supply Michel Transport Medium provides a stable medium for transport of fresh unfixed tissues, such as renal, rectal, lymph node, skin and oral mucosa biopsies, which will undergo subsequent frozen sectioning and immunofluorescence studies. Michel Transport Medium is not a fixative and does not have any fixative properties. Michel Transport Medium is not suitable for transporting cells for flow cytometry or tissues used for fluorescent in-situ hybridization (FISH) studies. Also available in pre-filled Michel Transport Medium Vials. Shelf Life is 1 year from date of manufacture. Store at 15-30°C.
Catalog No. NC0777939 Supplier Newcomer Supply Supplier No. 1242A
Newcomer Supply Michel Transport Medium provides a stable medium for transport of fresh unfixed tissues, such as renal, rectal, lymph node, skin and oral mucosa biopsies, which will undergo subsequent frozen sectioning and immunofluorescence studies. Michel Transport Medium is not a fixative and does not have any fixative properties. Michel Transport Medium is not suitable for transporting cells for flow cytometry or tissues used for fluorescent in-situ hybridization (FISH) studies. Also available in pre-filled Michel Transport Medium Vials. Shelf Life is 1 year from date of manufacture. Store at 15-30°C.